Novus Biologicals
Manufacturer Code:NBP227540
Catalog # NBP227540
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RPQIRLEREQLRT corresponding to internal region according to NP_001908.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: D-amino-acid oxidase; DAAO; DAAO D-amino-acid oxidase DAMOXOXDA DAO EC 1.4.3 EC 1.4.3.3 MGC35381
Gene Aliases: DAAO; DAMOX; DAO; OXDA
UniProt ID: (Human) P14920
Entrez Gene ID: (Human) 1610
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.