Invitrogen
Manufacturer Code:MA525684
Catalog # PIMA525684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2H3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 23-189 of DAND5 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cerberus 2; Cerberus 2 Cerberus like-2 CKTSF1B3 DAND5 Dante GREM3 CER2CRL2 cerberus-like 2 Cerberus-like protein 2 CERL2 Cerl-2 cerl-2 CKTSF1B3MGC126849 COCO Cysteine knot superfamily 1 BMP antagonist 3 DAN domain family member 5 DAN domain family member 5 DANTE DTE FLJ38607 GREM3cerberus 2 gremlin-3 Gremlin-3 SP1; cerberus-like 2; Cerberus-like protein 2; Cerl-2; Cysteine knot superfamily 1, BMP antagonist 3; DAN domain family member 5; DAN domain family member 5, BMP antagonist; DAN domain family, member 5; Gremlin-3
Gene Aliases: CER2; CERL2; CKTSF1B3; COCO; CRL2; DAND5; DANTE; GREM3; SP1
UniProt ID: (Human) Q8N907
Entrez Gene ID: (Human) 199699
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.