R&D Systems
Manufacturer Code:MAB7551SP
Catalog # MAB7551SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 123202 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse DAN Ala17-Asp178 Accession # Q61477.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: DAN domain family member 1; DAN domain family member 1 DAND1 MGC8972 NBL1 neuroblastoma suppressor of tumorigenicity 1 neuroblastoma suppression of tumorigenicity 1 Protein N03 suppression of tumorigenicity 1 Zinc finger protein DAN; differential screening-selected gene aberrant in neuroblastoma; neuroblastoma candidate region, suppression of tumorigenicity 1; Neuroblastoma suppressor of tumorigenicity 1; Protein N03; Zinc finger protein DAN
Gene Aliases: D1S1733E; DAN; DAND1; NB; NBL1; NO3
UniProt ID: (Human) P41271
Entrez Gene ID: (Human) 4681
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.