Novus Biologicals
Manufacturer Code:NBP15682220UL
Catalog # NBP15682220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to DALRD3(DALR anticodon binding domain containing 3) The peptide sequence was selected from the middle region of DALRD3. Peptide sequence TEIFGVLSVATIKFEMLSTAPQSQLFLALADSSISTKGTKSGTFVMYNCA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DALR anticodon binding domain containing 3 DALR anticodon-binding domain-containing protein 3 FLJ10496; DALR anticodon-binding domain-containing protein 3
Gene Aliases: DALRD3
UniProt ID: (Human) Q5D0E6
Entrez Gene ID: (Human) 55152
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.