Invitrogen
Manufacturer Code:PA5104535
Catalog # PIPA5104535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DAK corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent dihydroxyacetone kinase; ATP-dependent dihydroxyacetone kinase BC021917 Bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing) Dak DHA kinase Dha kinase/FMN cyclase dihydroxyacetone kinase 2 homolog dihydroxyacetone kinase 2 homolog (S. cerevisiae) DKFZP586B1621 FAD-AMP lyase (cyclic FMN forming) FAD-AMP lyase (cyclizing) FAD-AMP lyase cyclic FMN forming FAD-AMP lyase cyclizing FMN cyclase Glycerone kinase MGC5621 NET45 RGD1311026 testis tissue sperm-binding protein Li 84P TKFC Triokinase triokinase and FMN cyclase triokinase FMN cyclase triokinase/FMN cyclase Triose kinase; Bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing); DHA kinase; Dha kinase/FMN cyclase; dihydroxyacetone kinase 2 homolog; FAD-AMP lyase (cyclic FMN forming); FAD-AMP lyase (cyclizing); FAD-AMP lyase cyclic FMN forming; FAD-AMP lyase cyclizing; FMN cyclase; Glycerone kinase; testis tissue sperm-binding protein Li 84P; Triokinase; Triokinase/FMN cyclase; Triose kinase
Gene Aliases: BC021917; DAK; NET45; RGD1311026; TKFC
UniProt ID: (Rat) Q4KLZ6, (Human) Q3LXA3, (Mouse) Q8VC30
Entrez Gene ID: (Rat) 361730, (Human) 26007, (Mouse) 225913
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.