Invitrogen
Manufacturer Code:PA523765
Catalog # PIPA523765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 317-345 amino acids from the central region of human DAGLA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C11orf11 DAGL(ALPHA) DAGLALPHA NSDDR DGL-alpha neural stem cell-derived dendrite regulator sn1-specific diacylglycerol lipase alpha; DAGL-alpha; DGL-alpha; Diacylglycerol lipase-alpha; Neural stem cell-derived dendrite regulator; Sn1-specific diacylglycerol lipase alpha
Gene Aliases: C11orf11; DAGL(ALPHA); DAGLA; DAGLALPHA; KIAA0659; NSDDR
UniProt ID: (Human) Q9Y4D2, (Mouse) Q6WQJ1
Entrez Gene ID: (Human) 747, (Mouse) 269060
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.