Invitrogen
Manufacturer Code:PA5112820
Catalog # PIPA5112820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit DAD1 protein (1-113AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI323713 Dad1 DAD-1 Defender against cell death 1 Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit DAD1 Oligosaccharyl transferase subunit DAD1 oligosaccharyltransferase 2 homolog oligosaccharyltransferase subunit 2 (non-catalytic) OST2; DAD-1; Defender against cell death 1; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit DAD1; Oligosaccharyl transferase subunit DAD1; oligosaccharyltransferase 2 homolog; oligosaccharyltransferase subunit 2 (non-catalytic)
Gene Aliases: DAD1; OST2
UniProt ID: (Human) P61803
Entrez Gene ID: (Human) 1603
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.