Abnova
Manufacturer Code:PAB16563
Catalog # 89-118-433
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 12 amino acids of human DACT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DACT2.
For Research Use Only
Protein Aliases: bA503C24.7; C6orf116; Dapper antagonist of catenin 2; Dapper homolog 2; dapper homolog 2, antagonist of beta-catenin; dapper, antagonist of beta-catenin, homolog 2; DAPPER2; dishevelled-binding antagonist of beta-catenin 2; DPR2; FLJ31232; MGC133141; MGC133142; RP11-503C24.7
Gene Aliases: bA503C24.7; C6orf116; DACT2; DAPPER2; DPR2; PP13671
UniProt ID: (Human) Q5SW24
Entrez Gene ID: (Human) 168002
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.