OriGene
Manufacturer Code:TA809156
Catalog # 50-168-2607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI13H4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 289-555 of human DACT1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Dapper antagonist of catenin 1; DAPPER DAPPER1 DPR1 FRODO HDPR1 TBS2 THYEX3; Dapper homolog 1; dapper, antagonist of beta-catenin, homolog 1; dishevelled-binding antagonist of beta-catenin 1; hDPR1; Hepatocellular carcinoma novel gene 3 protein; heptacellular carcinoma novel gene 3
Gene Aliases: DACT1; DAPPER; DAPPER1; DPR1; FRODO; HDPR1; HNG3; THYEX3
UniProt ID: (Human) Q9NYF0
Entrez Gene ID: (Human) 51339
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.