Invitrogen
Manufacturer Code:PA551796
Catalog # PIPA551796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DACT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dapper antagonist of catenin 1; DAPPER DAPPER1 DPR1 FRODO HDPR1 THYEX3 dapper homolog 1 hepatocellular carcinoma novel gene 3 protein heptacellular carcinoma novel gene 3; Dapper homolog 1; dapper, antagonist of beta-catenin, homolog 1; dishevelled-binding antagonist of beta-catenin 1; hDPR1; Hepatocellular carcinoma novel gene 3 protein; heptacellular carcinoma novel gene 3
Gene Aliases: DACT1; DAPPER; DAPPER1; DPR1; FRODO; HDPR1; HNG3; THYEX3
UniProt ID: (Human) Q9NYF0
Entrez Gene ID: (Human) 51339
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.