Invitrogen
Manufacturer Code:PA5114208
Catalog # PIPA5114208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: LSLVQKKTHP VRTNKPRTSV NADPTKGLLR NGSVCVRAPG GVSQGNSVNL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4921528D17Rik AI115603 DACT1 dact1.S dact1-a dact1-b DAPPER dapper 1 dapper 1-A Dapper antagonist of catenin 1 Dapper homolog 1 dapper homolog 1 antagonist of beta-catenin dapper homolog 1 antagonist of beta-catenin (xenopus) dapper antagonist of beta-catenin dapper antagonist of beta-catenin homolog 1 dapper antagonist of beta-catenin homolog 1 (Xenopus laevis) DAPPER1 dapper1b dishevelled binding antagonist of beta catenin 1 dishevelled-binding antagonist of beta-catenin 1 dishevelled-binding antagonist of beta-catenin 1 S homeolog dishevelled-interacting protein dpr DPR1 Frd1 FRODO frodo 1 Frodo homolog Frodo1 functional regulator of dsh in ontogenesis hDPR1 Hepatocellular carcinoma novel gene 3 protein heptacellular carcinoma novel gene 3 HNG3 MDpr1 MTNG3 RGD1564008 signaling protein Thyex3 thymus expressed gene 3 Thymus-expressed novel gene 3 protein XDpr XDpr1b XELAEV_18041122mg; Dapper antagonist of catenin 1; Dapper homolog 1; dapper, antagonist of beta-catenin, homolog 1; dishevelled-binding antagonist of beta-catenin 1; hDPR1; Hepatocellular carcinoma novel gene 3 protein; heptacellular carcinoma novel gene 3
Gene Aliases: DACT1; DAPPER; DAPPER1; DPR1; FRODO; HDPR1; HNG3; THYEX3
UniProt ID: (Human) Q9NYF0
Entrez Gene ID: (Human) 51339
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.