Invitrogen
Manufacturer Code:PA5112090
Catalog # PIPA5112090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the center region of the Human DACH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430028N04Rik Dach2 dachshund 2 (Drosophila) dachshund family transcription factor 2 Dachshund homolog 2 FLJ31391 MGC138545 RP11-345E19.1; Dach2; Dachshund homolog 2
Gene Aliases: DACH2
UniProt ID: (Human) Q96NX9
Entrez Gene ID: (Human) 117154
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.