Proteintech
Manufacturer Code:252191AP150UL
Catalog # 50-173-4794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DACH2 Fusion Protein Ag18403 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dach2; DACH2 Dachshund homolog 2; Dachshund homolog 2
Gene Aliases: 9430028N04Rik; DACH2
UniProt ID: (Human) Q96NX9, (Mouse) Q925Q8
Entrez Gene ID: (Human) 117154, (Mouse) 93837
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.