Novus Biologicals
Manufacturer Code:NBP100136
Catalog # NBP100136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence SPVENTPQNNECK corresponding to internal region according to NP_542937.1 NP_542938.1 NP_004383.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: dac homolog; Dach1; Dach1 DACHdachshund homolog 1 dachshund homolog (Drosophila) dachshund homolog 1 (Drosophila) FLJ10138; Dachshund homolog 1
Gene Aliases: DACH; DACH1
UniProt ID: (Human) Q9UI36
Entrez Gene ID: (Human) 1602
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.