Proteintech
Manufacturer Code:109141AP150UL
Catalog # 50-556-617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DACH1 Fusion Protein Ag1354 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dac homolog; DACH DACH1 Dachshund homolog 1 Dachshund-1; Dach1; dachshund family transcription factor 1; Dachshund homolog 1
Gene Aliases: Dac; DACH; DACH1
UniProt ID: (Human) Q9UI36, (Mouse) Q9QYB2
Entrez Gene ID: (Human) 1602, (Rat) 306096, (Mouse) 13134
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.