R&D Systems
Manufacturer Code:MAB8064
Catalog # MAB8064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 883216 |
Immunogen | <EM>E. coli</EM>-derived recombinant human DAB2 Lys630-Ala770 Accession # P98082 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adaptor molecule disabled-2; Dab, mitogen-responsive phosphoprotein, homolog 2; Differentially expressed in ovarian carcinoma 2; Differentially-expressed protein 2; Differentially-expressed protein 2 disabled (Drosophila) homolog 2 (mitogen-responsive phosphoprotein) disabled homolog 2 mitogen-responsive phosphoprotein (Drosophila) Disabled-2 DOC2 DOC-2DOC2disabled homolog 2 FLJ26626 mitogen-responsive phosphoprotein; Disabled homolog 2; disabled homolog 2, mitogen-responsive phosphoprotein; DOC-2
Gene Aliases: DAB2; DOC-2; DOC2
UniProt ID: (Human) P98082
Entrez Gene ID: (Human) 1601
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.