Invitrogen
Manufacturer Code:PA587830
Catalog # PIPA587830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-185 of human DAB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dab reelin signal transducer 1; Dab, reelin signal transducer, homolog 1; Disabled homolog 1; yotari
Gene Aliases: AI956902; C630028C02Rik; DAB1; scm; scr; scrambler; yot
UniProt ID: (Human) O75553, (Mouse) P97318
Entrez Gene ID: (Human) 1600, (Mouse) 13131
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.