Invitrogen
Manufacturer Code:PA586617
Catalog # PIPA586617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 201-250 of Human Dab1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dab reelin signal transducer 1; Dab, reelin signal transducer, homolog 1; Disabled homolog 1; Disabled homolog 1 DAB1; yotari
Gene Aliases: AI956902; C630028C02Rik; DAB1; scm; scr; scrambler; yot
UniProt ID: (Human) O75553, (Rat) Q8CJH2, (Mouse) P97318
Entrez Gene ID: (Human) 1600, (Rat) 266729, (Mouse) 13131
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.