Invitrogen
Manufacturer Code:PA585748
Catalog # PIPA585748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of mouse Dab1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI956902 antibody C630028C02Rik antibody disabled 1 antibody scm antibody scr antibody scrambler antibody yot antibody; Disabled homolog 1; yotari
Gene Aliases: AI956902; C630028C02Rik; Dab1; scm; scr; scrambler; yot
UniProt ID: (Rat) Q8CJH2, (Mouse) P97318
Entrez Gene ID: (Rat) 266729, (Mouse) 13131
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.