Invitrogen
Manufacturer Code:PA523222
Catalog # PIPA523222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (18 FRNNDHPEITYRLRNDSN 35) of DAAM1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Disheveled-associated activator of morphogenesis 1
Gene Aliases: DAAM1; KIAA0666
UniProt ID: (Human) Q9Y4D1, (Mouse) Q8BPM0
Entrez Gene ID: (Dog) 490714, (Bovine) 513742, (Human) 23002, (Horse) 100051203, (Mouse) 208846, (Rat) 314212
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.