Invitrogen
Manufacturer Code:PA570020
Catalog # PIPA570020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human D9Ertd402e |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T cell activation inhibitor, mitochondrial; T-cell activation inhibitor, mitochondrial; TCAIM D9Ertd402e Toag1; TOAG-1; Tolerance associated gene-1 protein; tolerance-associated gene 1; Tolerance-associated gene-1
Gene Aliases: C22H3orf23; C3orf23; D9Ertd402e; Gm1129; RGD1560717; TCAIM; TOAG-1; TOAG1
UniProt ID: (Human) Q8N3R3, (Mouse) Q66JZ4, (Rat) P0DKR2
Entrez Gene ID: (Bovine) 515010, (Horse) 100054832, (Dog) 485567, (Rabbit) 100342706, (Human) 285343, (Mouse) 382117, (Rat) 363169, (Guinea pig) 100714703, (Zebrafish) 559258
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.