Invitrogen
Manufacturer Code:MA537614
Catalog # PIMA537614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 625CT1.5.4 |
Immunogen | KLH conjugated synthetic peptide between 365-391 amino acids from the Central region of human DRD4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AW125663 D(2C) dopamine receptor D(4) dopamine receptor D4DR D4R D4RA Dopamine 4 receptor Dopamine D4 receptor dopamine receptor 4 dopamine receptor D4 Drd4 Drd-4 seven transmembrane helix receptor; D(2C) dopamine receptor; D(4) dopamine receptor; Dopamine D4 receptor; dopamine receptor 4; seven transmembrane helix receptor
Gene Aliases: AW125663; D4DR; D4R; Drd-4; DRD4
UniProt ID: (Human) P21917, (Mouse) P51436
Entrez Gene ID: (Human) 1815, (Mouse) 13491
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.