Invitrogen
Manufacturer Code:PA577391
Catalog # PIPA577391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RRWEAARHTKLHSR corresponding to amino acid residues 215-228 of rat D4 Dopamine receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(2C) dopamine receptor; D(4) dopamine receptor; Dopamine D4 receptor; dopamine receptor 4; DRD4
Gene Aliases: AW125663; D4R; D4RA; Drd-4; Drd4
UniProt ID: (Mouse) P51436
Entrez Gene ID: (Rat) 25432, (Mouse) 13491
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.