Invitrogen
Manufacturer Code:PA533483
Catalog # PIPA533483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from C-terminus of human DRD4 / Dopamine Receptor D4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(2C) dopamine receptor; D(4) dopamine receptor; Dopamine D4 receptor; DRD4 D(4) dopamine receptor D4DR D(2C) dopamine receptor D4 dopamine receptor Dopamine receptor D4 Dopamine 4 receptor Dopamine D4 receptor; seven transmembrane helix receptor
Gene Aliases: D4DR; DRD4
UniProt ID: (Human) P21917
Entrez Gene ID: (Human) 1815
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.