Proteintech
Manufacturer Code:138951AP150UL
Catalog # 50-555-233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | D2HGDH Fusion Protein Ag4857 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-2-hydroxyglutarate dehydrogenase, mitochondrial; D2HGD D2HGDH; D2hgdh {ECO:0000250|UniProtKB:Q8N465}
Gene Aliases: AA408776; AA408778; AI325464; D2HGD; D2HGDH; RGD1307976
UniProt ID: (Human) Q8N465, (Mouse) Q8CIM3, (Rat) P84850
Entrez Gene ID: (Human) 728294, (Mouse) 98314, (Rat) 301624
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.