Abnova
Manufacturer Code:H00008528A01
Catalog # 89-004-393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | DDO (NP_003640 270 a.a. ~ 369 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant DDO.
For Research Use Only
Protein Aliases: aspartic oxidase; D-aspartate oxidase; D-aspartate oxidase, DDO; DASOX; DDO-1; DDO-2; FLJ45203
Gene Aliases: DASOX; DDO; DDO-1; DDO-2
UniProt ID: (Human) Q99489
Entrez Gene ID: (Human) 8528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.