R&D Systems
Manufacturer Code:MAB6174SP
Catalog # MAB6174SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 682516 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Cytosolic Sulfotransferase 2B1/SULT2B1 Asp2-Ser365 Accession # O00204 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alcohol sulfotransferase; Alcohol sulfotransferase EC 2.8.2 EC 2.8.2.2 HSST2 HSST2sulfotransferase family cytosolic 2B member 1 Hydroxysteroid sulfotransferase 2 ST2B1 Sulfotransferase 2B1 sulfotransferase family cytosolic 2B member 1 SULT2B1; Hydroxysteroid sulfotransferase 2; ST2B1; Sulfotransferase 2B1; Sulfotransferase family 2B member 1; Sulfotransferase family cytosolic 2B member 1; sulfotransferase family, cytosolic, 2B, member 1
Gene Aliases: HSST2; SULT2B1
UniProt ID: (Human) O00204
Entrez Gene ID: (Human) 6820
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.