Novus Biologicals
Manufacturer Code:NBP289869AF532
Catalog # NB068172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 11 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Cytosolic Sulfotransferase 1E1/SULT1E1 (NP_005411.1 Asn2-Ile294). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.8.2 EST-1 ESTMGC34459 estrone sulfotransferase ST1E1 STEestrogen sulfotransferase Sulfotransferase 1E1 sulfotransferase family 1E estrogen-preferring member 1 Sulfotransferase estrogen-preferringEC 2.8.2.4; EST-1; Estrogen sulfotransferase; estrone sulfotransferase; ST1E1; Sulfotransferase 1E1; sulfotransferase family 1E, estrogen-preferring, member 1; Sulfotransferase, estrogen-preferring
Gene Aliases: EST; EST-1; ST1E1; STE; SULT1E1
UniProt ID: (Human) P49888
Entrez Gene ID: (Human) 6783
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.