R&D Systems
Manufacturer Code:MAB5959
Catalog # MAB5959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 724429 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Cytosolic Sulfotransferase 1B1/SULT1B1 Leu2-Ile296 Accession # O43704 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.8.2 EC 2.8.2.- EC 2.8.2.1 EC 2.8.2.4 MGC13356 ST1B1 ST1B2 ST1B2sulfotransferase family cytosolic 1B member 1 Sulfotransferase 1B1 Sulfotransferase 1B2 sulfotransferase family cytosolic 1B member 1 SULT1B1 SULT1B2 Thyroid hormone sulfotransferase; ST1B1; ST1B2; sulfotransferase 1B1; Sulfotransferase 1B2; Sulfotransferase family cytosolic 1B member 1; sulfotransferase family, cytosolic, 1B, member 1; Thyroid hormone sulfotransferase
Gene Aliases: ST1B1; ST1B2; SULT1B1; SULT1B2
UniProt ID: (Human) O43704
Entrez Gene ID: (Human) 27284
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.