Novus Biologicals
Manufacturer Code:NBP289950JF646
Catalog # NB099910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 34 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Cytosolic Sulfotransferase 1A1/SULT1A1 (P50225-1 Glu2-Leu295). |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Aryl sulfotransferase 1; Aryl sulfotransferase 1 EC 2.8.2 EC 2.8.2.1 HAST1/HAST2 Phenol sulfotransferase 1 Phenol-sulfating phenol sulfotransferase 1 P-PST P-PST 1 PST ST1A1 ST1A3 STP1MGC5163 STPMGC131921 sulfotransferase 1A1 sulfotransferase family cytosolic 1A phenol-preferring member 1 Thermostable phenol sulfotransferase thermostable phenol sulfotransferase1 ts-PST TSPST1; HAST1/HAST2; P-PST 1; Phenol sulfotransferase 1; Phenol-sulfating phenol sulfotransferase 1; ST1A1; ST1A3; Sulfotransferase 1A1; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1; Thermostable phenol sulfotransferase; thermostable phenol sulfotransferase1; Ts-PST
Gene Aliases: HAST1/HAST2; OK/SW-cl.88; P-PST; PST; ST1A1; ST1A3; STP; STP1; SULT1A1; TSPST1
UniProt ID: (Human) P50225
Entrez Gene ID: (Human) 6817
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.