Novus Biologicals
Manufacturer Code:NBP276425
Catalog # NBP276425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PAN-CK (Cocktail) |
Immunogen | This Cytokeratin pan Antibody (PAN-CK (Cocktail)) was developed against human epidermal keratin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: 67 kDa cytokeratin; CK-1; cytokeratin 1; Cytokeratin-1; epidermolytic hyperkeratosis 1; Hair alpha protein; K1; keratin 1, type II; Keratin, type II cytoskeletal 1; Keratin-1; Type-II keratin Kb1
Gene Aliases: CK1; EHK; EHK1; EPPK; K1; KRT1; KRT1A; KRTA; NEPPK
UniProt ID: (Human) P04264
Entrez Gene ID: (Human) 3848
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.