Novus Biologicals
Manufacturer Code:NBP250221AF532
Catalog # NP20221A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | K92 |
Immunogen | This Cytokeratin pan Antibody (K92) was developed against hair keratin. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 67 kDa cytokeratin; CK-1; cytokeratin 1; Cytokeratin-1; epidermolytic hyperkeratosis 1; Hair alpha protein; K1; keratin 1, type II; Keratin, type II cytoskeletal 1; Keratin-1; Type-II keratin Kb1
Gene Aliases: CK1; EHK; EHK1; EPPK; K1; KRT1; KRT1A; KRTA; NEPPK
UniProt ID: (Human) P04264
Entrez Gene ID: (Human) 3848
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.