Invitrogen
Manufacturer Code:MA513203
Catalog # PIMA513203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | PAN-CK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Antibody Cocktail
For Research Use Only
Protein Aliases: 40-kDa keratin intermediate filament; 47 kDa cytokeratin; 56 kDa cytokeratin; 57kd keratin; 57kDa keratin; 58 kDa cytokeratin; 60-kDa keratin; Cell proliferation-inducing gene 46 protein; CK-10; CK-13; CK-14; CK-18; CK-19; CK-4; CK-5; CK-6A; CK-6D; CK-7; CK-8; cytokeratin 10; cytokeratin 13; cytokeratin 14; cytokeratin 18; cytokeratin 19; cytokeratin 4; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; Cytokeratin 7; cytokeratin 8; Cytokeratin endo A; Cytokeratin endo B; Cytokeratin-10; Cytokeratin-13; Cytokeratin-14; Cytokeratin-18; Cytokeratin-19; Cytokeratin-4; Cytokeratin-5; Cytokeratin-6A; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; Cytokeratin-7; Cytokeratin-8; Cytoskeletal 57 kDa keratin; Endo B; epidermal keratin 10; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolysis bullosa simplex, Dowling-Meara, Koebner; Hom s 5; intermediate filament protein; K10; K13; K14; K18; K19; K4; K5; K6A; K7; K8; kamp-keratin derived antimicrobial peptide; KDAMP; keratin 10, type I; keratin 13, type I; keratin 14, type I; keratin 18, type I; keratin 19, type I; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6A, , type II; keratin 6A, type II; keratin 7, type II; keratin 8, type II; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 13; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 2, basic, gene 4; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, basic, gene 7; keratin complex 2, basic, gene 8; keratin complex 2, gene 6a; Keratin D; keratin K6h; keratin, 55K type II cytoskeletal; keratin, epidermal type II, K6A; keratin, simple epithelial type I, K7; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 13; Keratin, type I cytoskeletal 14; Keratin, type I cytoskeletal 18; Keratin, type I cytoskeletal 19; Keratin, type I cytoskeletal 59 kDa; keratin, type I, 40-kd; Keratin, type II cytoskeletal 4; Keratin, type II cytoskeletal 5; Keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 7; Keratin, type II cytoskeletal 8; Keratin-10; Keratin-13; Keratin-14; Keratin-18; Keratin-19; Keratin-4; Keratin-5; Keratin-6-alpha; Keratin-6A; keratin-6C; Keratin-7; Keratin-8; krt5; krt7; mK6-alpha; pankeratin; Sarcolectin; suprabasal cytokeratin 10; tuftelin interacting protein 8; type II mesothelial keratin K7; type-II keratin Kb12; Type-II keratin Kb4; Type-II keratin Kb5; Type-II keratin Kb6; Type-II keratin Kb7; Type-II keratin Kb8
Gene Aliases: 2310016L08Rik; 3300001P10Rik; AA960620; AI626930; AI663979; AL022697; AU019895; AW108092; AW146334; BCIE; BIE; CARD2; CK-18; CK-4; CK-6C; CK-6E; CK-8; CK10; CK13; CK14; CK18; CK19; CK4; CK5; CK6A; CK6C; CK6D; CK7; CK8; CYK18; CYK4; CYK8; D130054E02Rik; D15Wsu77e; DDD; DDD1; EBS2; EBS3; EBS4; EHK; EndoA; EndoC; K10; K13; K14; K18; K19; K1C1; K1CS; K2C7; K2C8; K4; K5; K6A; K6C; K6D; K7; K8; Ker2; Kerd; KO; KPP; Krt-1.10; Krt-1.13; Krt-1.14; Krt-1.19; Krt-2.4; Krt-2.8; Krt1-10; Krt1-13; Krt1-14; Krt1-18; Krt1-19; KRT10; KRT13; KRT14; KRT18; KRT19; Krt2-4; Krt2-5; Krt2-6; Krt2-6a; Krt2-6c; Krt2-7; Krt2-8; KRT4; KRT5; KRT5A; Krt6; KRT6A; KRT6C; KRT6D; KRT7; KRT8; mK6[a]; MK6a; NFJ; PC3; PIG46; SCL; Tfip8; WSN1; WSN2
UniProt ID: (Human) P13645, (Human) P13646, (Human) P02533, (Human) P05783, (Human) P08727, (Human) P19013, (Human) P13647, (Human) P02538, (Human) P08729, (Human) P05787, (Mouse) Q9CXH6, (Mouse) P08730, (Mouse) Q61781, (Mouse) P05784, (Mouse) P19001, (Mouse) P07744, (Mouse) Q922U2, (Mouse) P50446, (Mouse) Q9DCV7, (Mouse) P11679
Entrez Gene ID: (Human) 3858, (Human) 3860, (Human) 3861, (Human) 3875, (Human) 3880, (Human) 3851, (Human) 3852, (Human) 3853, (Human) 3855, (Human) 3856, (Mouse) 16661, (Mouse) 16663, (Mouse) 16664, (Mouse) 16668, (Mouse) 16669, (Mouse) 16682, (Mouse) 110308, (Mouse) 16687, (Mouse) 110310, (Mouse) 16691
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.