Invitrogen
Manufacturer Code:MA126237
Catalog # PIMA126237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MNF116 |
Immunogen | Crude extract of splenic cells from a nude mouse engrafted with MCF-7 cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 39.1; 40-kDa keratin intermediate filament; 58 kDa cytokeratin; CK-17; CK-19; CK-5; CK-6A; CK-6D; CK-8; CK1 EHK EHK1 EPPK K1 KRT1A NEPPK pankeratin; cytokeratin 19; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; Cytokeratin-17; Cytokeratin-19; Cytokeratin-5; Cytokeratin-6A; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; Cytokeratin-8; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; Hom s 5; K17; K19; K5; K6A; K8; kamp-keratin derived antimicrobial peptide; KDAMP; keratin 17, type I; keratin 19, type I; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6A, , type II; keratin 6A, type II; keratin 8, type II; keratin K6h; keratin, epidermal type II, K6A; Keratin, type I cytoskeletal 17; Keratin, type I cytoskeletal 19; keratin, type I, 40-kd; Keratin, type II cytoskeletal 5; Keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 8; Keratin-17; Keratin-19; Keratin-5; Keratin-6A; keratin-6C; Keratin-8; type-II keratin Kb12; Type-II keratin Kb5; Type-II keratin Kb6; Type-II keratin Kb8
Gene Aliases: 39.1; CARD2; CK-17; CK-6C; CK-6E; CK-8; CK19; CK5; CK6A; CK6C; CK6D; CK8; CYK8; DDD; DDD1; EBS2; K17; K19; K1CS; K2C8; K5; K6A; K6C; K6D; K8; KO; KRT17; KRT19; KRT5; KRT5A; KRT6A; KRT6C; KRT6D; KRT8; PC; PC2; PC3; PCHC1
UniProt ID: (Human) Q04695, (Human) P08727, (Human) P13647, (Human) P02538, (Human) P05787
Entrez Gene ID: (Human) 3872, (Human) 3880, (Human) 3852, (Human) 3853, (Human) 3856
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.