Invitrogen
Manufacturer Code:MA512135
Catalog # PIMA512135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 34BetaE12 |
Immunogen | Solubilized keratin extracted from human stratum corneum |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 58 kDa cytokeratin; 67 kDa cytokeratin; CK-1; CK-10; CK-14; CK-5; cytokeratin 1; cytokeratin 10; cytokeratin 14; Cytokeratin-1; Cytokeratin-10; Cytokeratin-14; Cytokeratin-5; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis 1; Hair alpha protein; K1; K10; K14; K5; keratin 1, type II; keratin 10, type I; keratin 14, type I; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 14; Keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 5; Keratin-1; Keratin-10; Keratin-14; Keratin-5; Type-II keratin Kb1; Type-II keratin Kb5
Gene Aliases: BCIE; BIE; CK1; CK10; CK14; CK5; DDD; DDD1; EBS2; EBS3; EBS4; EHK; EHK1; EPPK; K1; K10; K14; K5; KPP; KRT1; KRT10; KRT14; KRT1A; KRT5; KRT5A; KRTA; NEPPK; NFJ
UniProt ID: (Human) P04264, (Human) P13645, (Human) P02533, (Human) P13647
Entrez Gene ID: (Human) 3848, (Rabbit) 100353415, (Horse) 100063302, (Rabbit) 100009029, (Human) 3858, (Human) 3861, (Horse) 100053489, (Rabbit) 100352658, (Horse) 100061626, (Human) 3852
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.