Invitrogen
Manufacturer Code:MA514428
Catalog # PIMA514428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TS1 |
Immunogen | Keratin preparation from a human carcinoma |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CK-8; CK-8 cytokeratin 8 cytokeratin-8 keratin type II cytoskeletal 8 keratin-8 type-II keratin Kb8; Cytokeratin-8; K8; keratin 8, type II; Keratin, type II cytoskeletal 8; Keratin-8; Type-II keratin Kb8
Gene Aliases: CARD2; CK-8; CK8; CYK8; K2C8; K8; KO; KRT8
UniProt ID: (Human) P05787
Entrez Gene ID: (Human) 3856
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.