Novus Biologicals
Manufacturer Code:NBP2448190.2MG
Catalog # NBP24481902
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | KRT7/760 + OV-TL12/30 |
Immunogen | Recombinant full-length human KRT7 protein (KRT7/760) OTN 11 ovarian carcinoma cell line (OV-TL12/30) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CK-7; CK7 CK-7 cytokeratin 7 Cytokeratin-7 K2C7 K7keratin 55K type II cytoskeletal keratin 7 keratin type II cytoskeletal 7 keratin-7 MGC129731 MGC3625 Sarcolectin SCLkeratin simple epithelial type I K7 type II mesothelial keratin K7 Type-II keratin Kb7; cytokeratin 7; Cytokeratin-7; K7; keratin 7, type II; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; Keratin, type II cytoskeletal 7; Keratin-7; Sarcolectin; type II mesothelial keratin K7; Type-II keratin Kb7
Gene Aliases: CK7; K2C7; K7; KRT7; SCL
UniProt ID: (Human) P08729
Entrez Gene ID: (Human) 3855
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.