Proteintech
Manufacturer Code:155391AP150UL
Catalog # 50-553-434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cytokeratin 7 Fusion Protein Ag7895 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK 7 CK7 Cytokeratin 7 K2C7 K7 keratin 7 KRT7 Sarcolectin SCL Type II keratin Kb7; CK-7; Cytokeratin 7; Cytokeratin-7; K7; keratin 7, type II; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; Keratin, type II cytoskeletal 7; Keratin-7; krt7; krt7 {ECO:0000250|UniProtKB:P08729}; Sarcolectin; type II keratin Kb7; type II mesothelial keratin K7; Type-II keratin Kb7
Gene Aliases: CK7; D15Wsu77e; K2C7; K7; Kb7; Krt2-7; KRT7; SCL
UniProt ID: (Human) P08729, (Mouse) Q9DCV7, (Rat) Q6IG12
Entrez Gene ID: (Human) 3855, (Mouse) 110310, (Rat) 300242
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.