Novus Biologicals
Manufacturer Code:NBP214924
Catalog # NBP214924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Cytokeratin 6B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK-6B; CK6B CK-6B cytokeratin 6B cytokeratin-6B K6B keratin 6B keratin epidermal type II K6B keratin type II cytoskeletal 6B keratin-6B keratin-like 1 (a type II keratin sequence) KRTL1 PC2 Type-II keratin Kb10; cytokeratin 6B; Cytokeratin-6B; K6B; keratin 6B, type II; keratin, epidermal, type II, K6B; Keratin, type II cytoskeletal 6B; Keratin-6B; keratin-like 1 (a type II keratin sequence); Type-II keratin Kb10
Gene Aliases: CK-6B; CK6B; K6B; KRT6B; KRTL1; PC2; PC4
UniProt ID: (Human) P04259
Entrez Gene ID: (Human) 3854
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.