Proteintech
Manufacturer Code:173911AP150UL
Catalog # 50-173-1546
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cytokeratin 6B Fusion Protein Ag11288 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK 6B CK6B Cytokeratin 6B K6B keratin 6B KRT6B KRTL1 PC2 Type II keratin Kb10; CK-6B; cytokeratin 6B; Cytokeratin-6B; K6B; keratin 6B, type II; keratin complex 2, basic, gene 6b; keratin, epidermal, type II, K6B; Keratin, type II cytoskeletal 6B; keratin-6-beta; Keratin-6B; keratin-like 1 (a type II keratin sequence); mK6-beta; Type-II keratin Kb10
Gene Aliases: BB144589; CK-6B; CK6B; K6B; Krt2-6b; KRT6B; KRTL1; mK6[b]; PC2; PC4
UniProt ID: (Human) P04259
Entrez Gene ID: (Human) 3854, (Mouse) 16688
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.