Invitrogen
Manufacturer Code:PA513684
Catalog # PIPA513684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 386-415 amino acids from the C-terminal region of human KRT4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 57kd keratin; 57kDa keratin; CK-4; cytokeratin 4; Cytokeratin-4; Cytoskeletal 57 kDa keratin; K4; keratin 4, type II; keratin complex 2, basic, gene 4; Keratin type II cytoskeletal 4 Cytokeratin-4 CK-4 Keratin-4 K4 CYK4 KRT4; Keratin, type II cytoskeletal 4; Keratin-4; Type-II keratin Kb4
Gene Aliases: AW108092; CK-4; CK4; CYK4; K4; Krt-2.4; Krt2-4; KRT4; WSN1
UniProt ID: (Human) P19013, (Mouse) P07744
Entrez Gene ID: (Human) 3851, (Mouse) 16682
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.