Invitrogen
Manufacturer Code:PA531775
Catalog # PIPA531775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 42 and 263 of Human Keratin 36 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HA6 KRTHA6 hHa6 K36 hair keratin type I Ha6 keratin hair acidic 6 keratin type I cuticular Ha6 keratin-36; Hair keratin, type I Ha6; K36; keratin 36, type I; keratin, hair, acidic, 6; Keratin, type I cuticular Ha6; Keratin-36
Gene Aliases: HA6; HHA6; HKA6; KRT36; KRTHA6
UniProt ID: (Human) O76013
Entrez Gene ID: (Human) 8689
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.