Proteintech
Manufacturer Code:173291AP150UL
Catalog # 50-173-1513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cytokeratin 20 Fusion Protein Ag11308 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK 20 CK20 Cytokeratin 20 K20 keratin 20 KRT20 KRT21 Protein IT; CK-20; CK-21; cytokeratin 20; cytokeratin 21; Cytokeratin-20; Cytokeratin-21; K20; keratin 20, type I; Keratin, type I cytoskeletal 20; Keratin-20; krt20; Protein IT
Gene Aliases: 9030623C06Rik; CD20; CK-20; CK20; K20; KRT20; KRT21
UniProt ID: (Human) P35900, (Mouse) Q9D312, (Rat) P25030
Entrez Gene ID: (Human) 54474, (Mouse) 66809, (Rat) 286912
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.