Novus Biologicals
Manufacturer Code:NBP216091
Catalog # NBP216091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Cytokeratin 2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK-2e; CK-2e Cytokeratin-2e epidermal ichthyosis bullosa of Siemens Epithelial keratin-2e K2e keratin 2 keratin 2A (epidermal ichthyosis bullosa of Siemens) keratin type II cytoskeletal 2 epidermal Keratin-2 epidermis keratin-2e KRT2AMGC116967 KRT2EMGC116968 KRTE Type-II keratin Kb2; Cytokeratin-2e; Epithelial keratin-2e; K2e; keratin 2, type II; Keratin, type II cytoskeletal 2 epidermal; Keratin-2 epidermis; Keratin-2e; Type-II keratin Kb2
Gene Aliases: CK-2e; K2e; KRT2; KRT2A; KRT2E; KRTE
UniProt ID: (Human) P35908
Entrez Gene ID: (Human) 3849
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.