Novus Biologicals
Manufacturer Code:NBP238536
Catalog # NBP238536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: SSRTSCRLSGGLGAGSCRLGSAGGL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 39.1; 39.1 CK-17 cytokeratin-17 K17 keratin 17 keratin type I cytoskeletal 17 keratin-17 PC PC2 PCHC1; CK-17; Cytokeratin-17; K17; keratin 17, type I; Keratin, type I cytoskeletal 17; Keratin-17
Gene Aliases: 39.1; CK-17; K17; KRT17; PC; PC2; PCHC1
UniProt ID: (Human) Q04695
Entrez Gene ID: (Human) 3872
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.