Invitrogen
Manufacturer Code:PA586163
Catalog # PIPA586163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 421-470 of Human Cytokeratin 16. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK-16; cytokeratin 16; Cytokeratin-16; focal non-epidermolytic palmoplantar keratoderma; K16; keratin 16, type I; keratin complex 1, acidic, gene 16; keratin intermediate filament 16a; keratin intermediate filament 16b; Keratin type I cytoskeletal 16 Cytokeratin-16 Cytokeratin16 CK-16 CK16 Keratin-16 Keratin 16 Keratin16 Short name&equalsK16 KRT16 KRT16A; Keratin, type I cytoskeletal 16; Keratin-16; type I keratin KA16
Gene Aliases: AI324768; CK16; FNEPPK; K16; K1CP; Ka16; Krt1-16; KRT16; KRT16A; NEPPK; PC1
UniProt ID: (Human) P08779, (Mouse) Q9Z2K1
Entrez Gene ID: (Human) 3868, (Mouse) 16666, (Rat) 303530
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.