Novus Biologicals
Manufacturer Code:NBP122777
Catalog # NBP122777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | KS-1A3 |
Immunogen | Cultured A431 cells from a human epidermoid carcinoma of the vulva. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CK-13; CK13 CK-13 cytokeratin 13 Cytokeratin-13 K13cytokeratin-13 keratin 13 keratin type I cytoskeletal 13 keratin-13 MGC161462 MGC3781; cytokeratin 13; Cytokeratin-13; K13; keratin 13, type I; Keratin, type I cytoskeletal 13; Keratin-13
Gene Aliases: CK13; K13; KRT13; WSN2
UniProt ID: (Human) P13646
Entrez Gene ID: (Human) 3860
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.