Proteintech
Manufacturer Code:666841IG150UL
Catalog # 50-173-7101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E10G2 |
Immunogen | Cytokeratin 13 Fusion Protein Ag0217 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CK 13 CK13 Cytokeratin 13 K13 keratin 13 KRT13; CK-13; cytokeratin 13; Cytokeratin-13; K13; keratin 13, type I; Keratin, type I cytoskeletal 13; Keratin-13; krt13 {ECO:0000250|UniProtKB:P13646}; Type I keratin Ka13
Gene Aliases: CK13; K13; Ka13; KRT13; WSN2
UniProt ID: (Human) P13646, (Rat) Q6IFV4
Entrez Gene ID: (Human) 3860, (Rat) 287699
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.