Invitrogen
Manufacturer Code:PA513663
Catalog # PIPA513663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 318-346 amino acids from the central region of human KRT10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK-10; cytokeratin 10; Cytokeratin-10; K10; keratin 10, type I; Keratin type I cytoskeletal 10 Cytokeratin-10 CK-10 Keratin-10 K10 KPP KRT10; Keratin, type I cytoskeletal 10; Keratin-10
Gene Aliases: BCIE; BIE; CK10; EHK; K10; KPP; KRT10
UniProt ID: (Human) P13645
Entrez Gene ID: (Human) 3858
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.