Invitrogen
Manufacturer Code:MA125510
Catalog # PIMA125510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CYT2-21 |
Immunogen | Recombit full length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARF exchange factor; ARF nucleotide binding site opener; ARF nucleotide-binding site opener; ARNO CTS18 CTS18.1 PSCD2 PSCD2L SEC7L Sec7p-L Sec7p-like; CLM2; CTC-273B12.8; Cytohesin-2; PH, SEC7 and coiled-coil domain-containing protein 2; pleckstrin homology, Sec7 and coiled-coil domains 2; pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2); pleckstrin homology, Sec7 and coiled/coil domains 2 (cytohesin-2); Protein ARNO; SEC7 homolog B
Gene Aliases: ARNO; CTS18; CTS18.1; CYTH2; PSCD2; PSCD2L; Sec7; Sec7B; SEC7L; Sec7p-L; Sec7p-like
UniProt ID: (Human) Q99418, (Rat) P63035
Entrez Gene ID: (Human) 9266, (Rat) 116692
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.